n-Butyllithium, 2.7M solution in toluene, AcroSeal™
n-Butyllithium, 2.7M solution in toluene, AcroSeal™
n-Butyllithium, 2.7M solution in toluene, AcroSeal™
n-Butyllithium, 2.7M solution in toluene, AcroSeal™
Thermo Scientific Chemicals

n-Butyllithium, 2.7M solution in toluene, AcroSeal™

CAS: 109-72-8 | C4H9Li | 64.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 378931000
Price (USD)
57.65
Online Exclusive
64.30
Save 6.65 (10%)
Each
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
57.65
Online Exclusive
64.30
Save 6.65 (10%)
Each
Add to cart
Chemical Identifiers
CAS109-72-8
IUPAC Namebutyllithium
Molecular FormulaC4H9Li
InChI KeyMZRVEZGGRBJDDB-UHFFFAOYSA-N
SMILES[Li]CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Active base2.62 to 2.75 M
Appearance (Color)Yellow to orange
Appearance (Form)Solution
AppearanceMay develop some turbidity or precipitate
Free base=<1.0 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection