Thermo Scientific™

HAAKE™ MARS™ iQ Rotational Rheometer

Catalog number: 379-0800
Have Questions?
Thermo Scientific™

HAAKE™ MARS™ iQ Rotational Rheometer

Catalog number: 379-0800
Have Questions?
Thermo Scientific™ HAAKE™ MARS™ iQ Rotational Rheometers ensure intuitive, reliable and flexible rheological measurements. Everything about this QC rheometer is designed to provide intuitive handling and fast, reliable results. A sophisticated touchscreen enables standard operating procedures to be run right from rheometer itself. “Connect Assist” functionality guides users for failure-free measurement.
 
Choose a HAAKE MARS iQ Rheometer with either the ball bearing or air bearing system to suit your lab. A broad range of accessories helps meet the ever-greater testing demands on QC labs.
 
HAAKE MARS iQ Rotational Rheometers are the replacements for the discontinued Thermo Scientific™ HAAKE™ RheoStress™ 1 Rheometer and Thermo Scientific™ HAAKE™ RotoVisco™ 1 Rotational Rheometer.
 
Catalog Number
379-0800
Unit Size
Each
Description
HAAKE MARS iQ Rheometer - ball-bearing
Price (USD)
Full specifications
Angular Resolution0.63 μrad
DescriptionHAAKE MARS iQ Rheometer - ball-bearing
Dimensions (L x W x H)480 x 390 x 670 mm
Frequency Range0.01 to 20 Hz
Lift Speed Range0.05 μm/sec. to 20 mm/sec.
Rotational Speed Range0.01 rpm to 2000 rpm
Temperature Range (Metric)-60°C to +400°C
Torque RangeMin. torque rotation: 0.2 mNm
Max. torque rotation: 125 mNm
Min. torque oscillation: 0.2 mNm
Max torque oscillation: 125 mNm
Torque resolution: 2.0 μNm
Velocity Range0.001 rad/s to 209.4 rad/s
Weight (Metric)57 kg
Unit SizeEach
Catalog Number
Unit Size
Each
Description
HAAKE MARS iQ Rheometer - air-bearing
Price (USD)
Full specifications
Angular Resolution0.63 μrad
DescriptionHAAKE MARS iQ Rheometer - air-bearing
Dimensions (L x W x H)480 x 390 x 670 mm
Frequency Range0.0001 to 100 Hz
Lift Speed Range0.05 μm/sec. to 20 mm/sec.
Normal Force(N): 0.01 to 50
Normal force resolution (N): 0.001
Rotational Speed Range0.001 to 2000 rpm
Temperature Range (Metric)-60°C to +400°C
Torque RangeMin. torque rotation: 0.001 mNm
Max. torque rotation: 150 mNm
Min. torque oscillation: 0.001 mNm
Max torque oscillation: 150 mNm
Torque resolution: 0.007 μNm
Velocity Range0.0001 to 209.4 rad/s
Weight (Metric)57 kg
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
379-0800Full specifications
EachHAAKE MARS iQ Rheometer - ball-bearingRequest A Quote
Angular Resolution0.63 μrad
DescriptionHAAKE MARS iQ Rheometer - ball-bearing
Dimensions (L x W x H)480 x 390 x 670 mm
Frequency Range0.01 to 20 Hz
Lift Speed Range0.05 μm/sec. to 20 mm/sec.
Rotational Speed Range0.01 rpm to 2000 rpm
Temperature Range (Metric)-60°C to +400°C
Torque RangeMin. torque rotation: 0.2 mNm
Max. torque rotation: 125 mNm
Min. torque oscillation: 0.2 mNm
Max torque oscillation: 125 mNm
Torque resolution: 2.0 μNm
Velocity Range0.001 rad/s to 209.4 rad/s
Weight (Metric)57 kg
Unit SizeEach
379-0700Full specifications
EachHAAKE MARS iQ Rheometer - air-bearingRequest A Quote
Angular Resolution0.63 μrad
DescriptionHAAKE MARS iQ Rheometer - air-bearing
Dimensions (L x W x H)480 x 390 x 670 mm
Frequency Range0.0001 to 100 Hz
Lift Speed Range0.05 μm/sec. to 20 mm/sec.
Normal Force(N): 0.01 to 50
Normal force resolution (N): 0.001
Rotational Speed Range0.001 to 2000 rpm
Temperature Range (Metric)-60°C to +400°C
Torque RangeMin. torque rotation: 0.001 mNm
Max. torque rotation: 150 mNm
Min. torque oscillation: 0.001 mNm
Max torque oscillation: 150 mNm
Torque resolution: 0.007 μNm
Velocity Range0.0001 to 209.4 rad/s
Weight (Metric)57 kg
Unit SizeEach
Showing 2 of 2

HAAKE MARS iQ Rheometers offer many advantages:

Intuitive Operations

  • Multifunctional 7’’ touchscreen for instrument operation and SOP execution right at your fingertips
  • ‘‘Assist‘‘ functionalities to simplify operation and prevent mistakes
  • ‘‘Connect Assist‘‘ quick coupling of measuring geometries and temperature modules with automatic recognition
  • ‘‘Color Assist‘‘ color-coded plugs for quick, correct selection of temperature modules
  • HAAKE™ RheoWin™ Rheology Software for extensive measuring flexibility with free updates (Optional 21 CFR part 11 module available)

Intelligent Design

  • Folded H frame concept for greater stiffness and ultra-precise lift control with a wide travel range
  • Durable material: high-performance mineral composite casting with high vibrational damping, minimal temperature expansion and high chemical resistance
  • Next generation of highly dynamic, powerful EC motor with a mechanical or air bearing. 

Individualized Configurations

  • Broad scope of temperature modules, measuring geometries and application-oriented measuring cells, e.g., pressure cells, building material measuring cell or tribology measuring cell
  • Sensitive normal force sensor measures axial forces in both directions for applications such as texture analysis
  • Lateral and bottom access for customized test requirements

Application solutions
The HAAKE MARS iQ Rheometers are well-suited for the rheological characterization of a broad range of samples from low-viscous or water-like up to stiff and paste-like semisolids:

  • Investigation of paints, coatings and inks: shear viscosity, thixotropy, yield stress, shear recovery, levelling and sagging behavior
  • Rheology and textural properties of foods and foodstuffs: shear viscosity, yield stress, tribology, texture analysis (axial bending, breaking, squeezing tests), phase transition  
  • Rheology of cosmetics and pharmaceuticals: viscosity, yield stress, viscoelastic properties, stability tests 
  • Rheology of polymer (e.g. polymer melts and solutions): viscoelasticity, phase transitions (glass transition, crystallization, melting), processing behavior (hot melt extrusion, molding, spinning), molecular weight, curing
  • Rheology of petrochemicals (crude oils, fracking liquids and drilling fluids): viscosity, yield stress, pressure dependent properties, stability tests, wax deposition, wax appearance temperature (WAT)
  • Dynamic shear rheometer (DSR) tests according to AASHTO, ASTM and DIN used for bitumen and asphalt
  • Construction and building material (cement paste and slurries): flow properties of suspensions with large particles

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text