Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store and track samples in Thermo Scientific™ Matrix™ 2D Barcoded Clear Polypropylene Open-Top Storage Tubes. Laser etching produces a permanent, high-contrast, 2D barcode offering maximum reliability and traceability of compound, biological and genomic samples. Offered in three sizes, 0.5mL, 0.75mL, and 1.4mL, these tubes can be sealed with Thermo Scientific SepraSeal septum caps and stored down to -80°C. Item number 3792 is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Capacity (Metric) | 1.4 mL |
Bottom Shape | V-bottom |
Material | Polypropylene |
Packaging | 5 Latch Racks of 96 Tubes |
Well Shape | V-bottom |
Barcode | 2D BarCoded |
Product Line | Matrix™ |
Sterility | Non-sterile |
Capacity (English) | 0.0473 fl oz. |
Fit for Purpose | PCR Ready |
Unit Size | Case of 960 |
Product Type | Storage Tube (Open Top) |
Capacity (Metric) | 1.4 mL |
Material | Polypropylene |
Packaging | 10 Latch Racks of 96 Tubes |
Well Shape | Flat |
Barcode | 2D barcode, TTPcomPOUND System |
Sterility | Non-sterile |
Capacity (English) | 0.0473 fl oz. |
Fit for Purpose | PCR Ready |
Unit Size | Each |
Product Type | Storage Tube (Open Top) |
Capacity (Metric) | 0.5 mL |
Bottom Shape | V-bottom |
Material | Polypropylene |
Packaging | 10 Latch Racks of 96 Tubes |
Well Shape | V-bottom |
Barcode | 2D BarCoded |
Sterility | Sterile |
Capacity (English) | 0.0169 fl oz. |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Each |
Product Type | Storage Tube (Open Top) |
Capacity (Metric) | 1.4 mL |
Material | Polypropylene |
Packaging | 10 Latch Racks of 96 Tubes |
Well Shape | Flat |
Barcode | 2D barcode, human readable |
Sterility | Non-sterile |
Capacity (English) | 0.0473 fl oz. |
Fit for Purpose | PCR Ready |
Unit Size | Each |
Capacity (Metric) | 1.4 mL |
Material | Polypropylene |
Packaging | Bulk, 1000 Tubes |
Well Shape | Flat |
Barcode | 2D barcoded w/human readable code |
Product Line | Matrix™ |
Sterility | Non-sterile |
Capacity (English) | 0.0473 fl oz. |
Fit for Purpose | PCR Ready |
Unit Size | Case of 1000 |
Product Type | Storage Tube (Open Top) |
Capacity (Metric) | 0.75 mL |
Bottom Shape | V-bottom |
Material | Polypropylene |
Packaging | Bulk, 1000 Tubes |
Well Shape | V-bottom |
Sterility | Non-sterile |
Capacity (English) | 0.0253 fl oz. |
Fit for Purpose | PCR Ready |
Unit Size | Case of 1000 |
Product Type | Storage Tube (Open Top) |
Capacity (Metric) | 0.75 mL |
Bottom Shape | V-bottom |
Material | Polypropylene |
Packaging | 10 Latch Racks of 96 Tubes |
Well Shape | V-bottom |
Sterility | Non-sterile |
Capacity (English) | 0.0253 fl oz. |
Fit for Purpose | PCR Ready |
Unit Size | Each |
Product Type | Storage Tube (Open Top) |
Capacity (Metric) | 1.4 mL |
Material | Polypropylene |
Packaging | 10 Latch Racks of 96 Tubes |
Well Shape | Flat |
Sterility | Sterile |
Capacity (English) | 0.0473 fl oz. |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Each |
Capacity (Metric) | 1.4 mL |
Material | Polypropylene |
Packaging | 5 Latch Racks of 96 Tubes |
Well Shape | Flat |
Barcode | 2D barcoded w/human readable code |
Product Line | Matrix™ |
Sterility | Non-sterile |
Capacity (English) | 0.0473 fl oz. |
Fit for Purpose | PCR Ready |
Unit Size | Case of 960 |
Capacity (Metric) | 1.4 mL |
Material | Polypropylene |
Packaging | 5 Latch Racks of 96 Tubes |
Well Shape | Flat |
Barcode | 2D barcoded w/human readable code |
Product Line | Matrix™ |
Sterility | Sterile |
Capacity (English) | 0.0473 fl oz. |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Sterility | Well Shape | Fit for Purpose | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
3791 | Case of 960 | 1.4 mL | Non-sterile | V-bottom | PCR Ready | 726.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
3719-11 | Each | 1.4 mL | Non-sterile | Flat | PCR Ready | 786.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
3725TS also known as 3725 | Each | 0.5 mL | Sterile | V-bottom | Sterile & PCR Ready | 784.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
3715-11 | Each | 1.4 mL | Non-sterile | Flat | PCR Ready | 805.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
3710 | Case of 1000 | 1.4 mL | Non-sterile | Flat | PCR Ready | 711.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
3730-11 | Case of 1000 | 0.75 mL | Non-sterile | V-bottom | PCR Ready | 726.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
3731-11 | Each | 0.75 mL | Non-sterile | V-bottom | PCR Ready | 729.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
3712-11 | Each | 1.4 mL | Sterile | Flat | Sterile & PCR Ready | 756.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
3711 | Case of 960 | 1.4 mL | Non-sterile | Flat | PCR Ready | 729.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
3712 | Each | 1.4 mL | Sterile | Flat | Sterile & PCR Ready | Request A Quote | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Secure Tracking:
Robust Storage Rack Design:
Stringent Quality Control:
Storage Tracking Solutions for Any Laboratory:
Recommended for:
Individual sample retrieval, instant sample identification, secure identification of archived samples, secure shipping of samples, storage of samples and tracking of samples.
Recycling Information:
Matrix tubes, tube racks and covers are made from polypropylene copolymer and are recyclable in communities where polypropylene is recycled. Please recycle only clean, dry components that have been decontaminated of any hazardous materials or residues. The orange nylon rack latches must be removed from the tube rack base and discarded before recycling the rack base. Tip: Orange latches can be removed with relative ease with the aid of pliers.
Matrix tubes are delivered in high graphic paper cartons. Please recycle the paper cartons and product inserts.