Thermo Scientific Chemicals

2,6-Difluorophenylacetone, 98%, Thermo Scientific Chemicals

Catalog number: 379150050
5 g, Each
Thermo Scientific Chemicals

2,6-Difluorophenylacetone, 98%, Thermo Scientific Chemicals

Catalog number: 379150050
5 g, Each
Quantity
Catalog number: 379150050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
101712-20-3
IUPAC Name
1-(2,6-difluorophenyl)propan-2-one
Molecular Formula
C9H8F2O
InChI Key
LAWHOFKPDMZDLJ-UHFFFAOYSA-N
SMILES
CC(=O)CC1=C(F)C=CC=C1F
GC
>=97.5 %
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4790 to 1.4820 (20°C, 589 nm)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text