Thermo Scientific Chemicals

Lithium hydroxide, 98%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 379181000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Lithium hydroxide, 98%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 379181000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 379181000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-65-2
IUPAC Name
lithium(1+) hydroxide
Molecular Formula
HLiO
InChI Key
WMFOQBRAJBCJND-UHFFFAOYSA-M
SMILES
[Li+].[OH-]
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
Titration with HCl
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text