Lithium hydroxide, 98%, pure, anhydrous
Lithium hydroxide, 98%, pure, anhydrous
Lithium hydroxide, 98%, pure, anhydrous
Lithium hydroxide, 98%, pure, anhydrous
Thermo Scientific Chemicals

Lithium hydroxide, 98%, pure, anhydrous

CAS: 1310-65-2 | HLiO | 23.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 379185000
Price (USD)
253.65
Special offer
Online exclusive
Ends: 26-Sep-2025
313.00
Save 59.35 (19%)
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
253.65
Special offer
Online exclusive
Ends: 26-Sep-2025
313.00
Save 59.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS1310-65-2
IUPAC Namelithium(1+) hydroxide
Molecular FormulaHLiO
InChI KeyWMFOQBRAJBCJND-UHFFFAOYSA-M
SMILES[Li+].[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HCl>=97.5 %
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link