Cobalt(II) oxide, Co >74%, -325 mesh, Thermo Scientific Chemicals
Cobalt(II) oxide, Co >74%, -325 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) oxide, Co >74%, -325 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 379200100
Price (USD)/ Each
58.65
Online exclusive
65.10 
Save 6.45 (10%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
58.65
Online exclusive
65.10 
Save 6.45 (10%)
Add to cart
Cobalt(II) oxide, Co >74%, -325 mesh, Thermo Scientific Chemicals
Catalog number379200100
Price (USD)/ Each
58.65
Online exclusive
65.10 
Save 6.45 (10%)
-
Add to cart
Chemical Identifiers
CAS1307-96-6
IUPAC Nameoxocobalt
Molecular FormulaCoO
InChI KeyIVMYJDGYRUAWML-UHFFFAOYSA-N
SMILESO=[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
Additional infoMay contain various amounts of Co(III)
Additional infooxides
Appearance (Form)Powder
Purity>=74.0 % Co
Appearance (Color)Dark grey to green-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only