Thermo Scientific Chemicals

Methyl α-bromophenylacetate, 97+%, Thermo Scientific Chemicals

Catalog number: 379300250
25 g, Each
Thermo Scientific Chemicals

Methyl α-bromophenylacetate, 97+%, Thermo Scientific Chemicals

Catalog number: 379300250
25 g, Each
Quantity
Catalog number: 379300250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3042-81-7
IUPAC Name
methyl 2-bromo-2-phenylacetate
Molecular Formula
C9H9BrO2
InChI Key
NHFBYYMNJUMVOT-UHFFFAOYNA-N
SMILES
COC(=O)C(Br)C1=CC=CC=C1
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.0 %
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text