Thermo Scientific Chemicals

Cerium(III) carbonate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 379400500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Cerium(III) carbonate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 379400500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 379400500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54451-25-1
IUPAC Name
dicerium(3+) tricarbonate
Molecular Formula
C3Ce2O9
InChI Key
GHLITDDQOMIBFS-UHFFFAOYSA-H
SMILES
[Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Appearance (Color)
White to off-white
Total trace metal impurities
=<0.1 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text