Thermo Scientific Chemicals

2,4-Dichloropyridine, 98%, Thermo Scientific Chemicals

Catalog number: 379470250
25 g, Each
Thermo Scientific Chemicals

2,4-Dichloropyridine, 98%, Thermo Scientific Chemicals

Catalog number: 379470250
25 g, Each
Quantity
Catalog number: 379470250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
26452-80-2
IUPAC Name
2,4-dichloropyridine
Molecular Formula
C5H3Cl2N
InChI Key
TYPVHTOETJVYIV-UHFFFAOYSA-N
SMILES
ClC1=CC=NC(Cl)=C1
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.5500 to 1.5540 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance
May darken on storage
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text