Thermo Scientific Chemicals

Tetraethylammonium perchlorate, 0.2M aqueous solution, Thermo Scientific Chemicals

Catalog number: 379805000
500 mL, Plastic bottle, Each
Thermo Scientific Chemicals

Tetraethylammonium perchlorate, 0.2M aqueous solution, Thermo Scientific Chemicals

Catalog number: 379805000
500 mL, Plastic bottle, Each
Quantity
Packaging
Catalog number: 379805000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2567-83-1, 7732-18-5
Appearance (Color)
Clear colorless
Appearance (Form)
Solution
Strength
0.2 M Exact strength on the certificate of analysis

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent, Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text