Thermo Scientific Chemicals

1-(2-Bromoethoxy)-4-chlorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: 379830050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1-(2-Bromoethoxy)-4-chlorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: 379830050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 379830050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2033-76-3
IUPAC Name
1-(2-bromoethoxy)-4-chlorobenzene
Molecular Formula
C8H8BrClO
InChI Key
YYFLBDSMQRWARK-UHFFFAOYSA-N
SMILES
ClC1=CC=C(OCCBr)C=C1
Appearance (Color)
White to almost white
Appearance (Form)
Crystalline powder or solid
Melting point
38.0°C to 42.0°C
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text