4-Methoxypyridine, 99%, Thermo Scientific Chemicals
4-Methoxypyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxypyridine, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number 379940050
Price (USD)/ Each
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-methoxypyridine
Molecular FormulaC6H7NO
InChI KeyXQABVLBGNWBWIV-UHFFFAOYSA-N
SMILESCOC1=CC=NC=C1
Molecular Weight (g/mol)109.13
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only