Thermo Scientific Chemicals

2-Bromophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 379990250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Bromophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 379990250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 379990250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
244205-40-1
IUPAC Name
(2-bromophenyl)boronic acid
Molecular Formula
C6H6BBrO2
InChI Key
PLVCYMZAEQRYHJ-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC=C1Br
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
HPLC
>=96.0 %
Titration with NaOH
96.0 to 110.0 %
Appearance (Form)
Powder or crystals and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text