Tin(IV) acetate, 98%, extra pure, Thermo Scientific Chemicals
Tin(IV) acetate, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(IV) acetate, 98%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 380050050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2800-96-6
IUPAC Nametin(4+) tetraacetate
Molecular FormulaC8H12O8Sn
InChI KeyYJGJRYWNNHUESM-UHFFFAOYSA-J
SMILES[Sn+4].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point231°C to 237°C
Appearance (Form)Crystalline powder
Gravimetry32.6 to 34.3 % Sn After ignition to oxide
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only