Sodium aluminum hydride, 93%, Thermo Scientific Chemicals
Sodium aluminum hydride, 93%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium aluminum hydride, 93%, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 g
Catalog number 380115000
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS13770-96-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey
Appearance (Form)Crystalline powder
Assay>=92 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only