Thermo Scientific Chemicals

Sodium hydroxide, for analysis, 50% solution in water, ACROS Organics™

Catalog number: 380210100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

Sodium hydroxide, for analysis, 50% solution in water, ACROS Organics™

Catalog number: 380210100
10 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 380210100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1310-73-2
IUPAC Name
sodium hydroxide
Molecular Formula
HNaO
InChI Key
HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES
[OH-].[Na+]
Appearance (Color)
Clear colorless
Iron (Fe)
=<0.0005 %
Sodium chloride (NaCl)
=<0.008 %
Appearance (Form)
Viscous solution
Titration with HCl
49 to 51 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text