Sodium hydroxide, for analysis, 50 wt% solution in water
Sodium hydroxide, for analysis, 50 wt% solution in water
Sodium hydroxide, for analysis, 50 wt% solution in water
Sodium hydroxide, for analysis, 50 wt% solution in water
Thermo Scientific Chemicals

Sodium hydroxide, for analysis, 50 wt% solution in water

CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
10 L
Packaging:
View all
Plastic bottle
Catalog number 380215000
Price (USD)
45.65
Online Exclusive
50.00
Save 4.35 (9%)
Each
Add to cart
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
45.65
Online Exclusive
50.00
Save 4.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Iron (Fe)=<0.0005 %
Sodium chloride (NaCl)=<0.008 %
Appearance (Form)Viscous solution
Titration with HCl49 to 51 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection