Thermo Scientific Chemicals

4-Bromophenylacetone, 97%, Thermo Scientific Chemicals

Catalog number: 380220050
5 g, Each
Thermo Scientific Chemicals

4-Bromophenylacetone, 97%, Thermo Scientific Chemicals

Catalog number: 380220050
5 g, Each
Quantity
Catalog number: 380220050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6186-22-7
IUPAC Name
methyl 2-{2-[N-(3,5-dimethylphenyl)benzenesulfonamido]acetamido}benzoate
Molecular Formula
C24H24N2O5S
InChI Key
CDUPPXGKUKTGLU-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Color)
Clear yellow
Appearance (Form)
Liquid after melting
Refractive index
1.5560 to 1.5590 (20°C, 589 nm)
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text