Specifications
Chemical Name or MaterialSodium phosphate dibasic
Melting Point250°C
CAS7558-79-4
ColorWhite
Quantity12 kg
Molecular FormulaNa2HPO4
SynonymDisodium hydrogen phosphate, Disodium hydrogen orthophosphate
Molecular Weight (g/mol)141.96
Percent Purity98.0-100.5%
GradeMulti-Compendial
PackagingPolyethylene pail, screw top
Unit SizeEach