Thermo Scientific Chemicals

Sodium diformamide, 98%, Thermo Scientific Chemicals

Catalog number: 380430050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium diformamide, 98%, Thermo Scientific Chemicals

Catalog number: 380430050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 380430050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
18197-26-7
IUPAC Name
sodium diformylazanide
Molecular Formula
C2H2NNaO2
InChI Key
QJXDSDLNUKLDBP-UHFFFAOYSA-M
SMILES
[Na+].O=C[N-]C=O
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with HCl
>=97.5 %
Appearance (Color)
Light beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text