Thermo Scientific Chemicals

6-Methoxy-3-pyridinecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 380570010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

6-Methoxy-3-pyridinecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 380570010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 380570010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
65873-72-5
IUPAC Name
6-methoxypyridine-3-carbaldehyde
Molecular Formula
C7H7NO2
InChI Key
CTAIEPPAOULMFY-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=O)C=N1
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Appearance (Color)
Off-white to light yellow
GC
>=96.0 %
Melting point
46°C to 54°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text