Thermo Scientific Chemicals

Butyl p-toluenesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 380595000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Butyl p-toluenesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 380595000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 380595000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
778-28-9
IUPAC Name
butyl 4-methylbenzene-1-sulfonate
Molecular Formula
C11H16O3S
InChI Key
QYJXDIUNDMRLAO-UHFFFAOYSA-N
SMILES
CCCCOS(=O)(=O)C1=CC=C(C)C=C1
Appearance (Color)
Clear colorless to yellow to brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.5030 to 1.5050 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text