Thermo Scientific Chemicals

Propyl p-toluenesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 380601000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Propyl p-toluenesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 380601000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 380601000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
599-91-7
IUPAC Name
propyl 4-methylbenzene-1-sulfonate
Molecular Formula
C10H14O3S
InChI Key
JTTWNTXHFYNETH-UHFFFAOYSA-N
SMILES
CCCOS(=O)(=O)C1=CC=C(C)C=C1
Refractive index
1.5065 to 1.5085 (20°C, 589 nm)
Infrared spectrum
Conforms
GC
>=98.0 %
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text