Thermo Scientific Chemicals

4-Piperidinemethanol, 98%, Thermo Scientific Chemicals

Catalog number: 380630250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Piperidinemethanol, 98%, Thermo Scientific Chemicals

Catalog number: 380630250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 380630250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6457-49-4
IUPAC Name
4-(hydroxymethyl)piperidin-1-ium
Molecular Formula
C6H14NO
InChI Key
XBXHCBLBYQEYTI-UHFFFAOYSA-O
SMILES
OCC1CC[NH2+]CC1
GC
>=97.5 %
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder and/or chunks or solid
Melting point
55°C to 62°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text