Starch, pure, 1 wt.% aqueous solution; stabilized w. 100ppb HgI2
Starch, pure, 1 wt.% aqueous solution; stabilized w. 100ppb HgI2
Starch, pure, 1 wt.% aqueous solution; stabilized w. 100ppb HgI2
Starch, pure, 1 wt.% aqueous solution; stabilized w. 100ppb HgI2
Starch, pure, 1 wt.% aqueous solution; stabilized w. 100ppb HgI2
Thermo Scientific Chemicals

Starch, pure, 1 wt.% aqueous solution; stabilized w. 100ppb HgI2

CAS: 9005-84-9 | (C6H10O5)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Plastic bottle
Catalog number 380641000
Price (USD)
-
Quantity:
100 mL
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9005-84-9
IUPAC Name(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyGUBGYTABKSRVRQ-ASMJPISFSA-N
SMILESOC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
SensitivityPasses test
Appearance (Form)Clear to slightly hazy solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link