Thermo Scientific Chemicals

Starch, Thermo Scientific Chemicals

Catalog number: 380645000
500 mL, Plastic bottle, Each
Thermo Scientific Chemicals

Starch, Thermo Scientific Chemicals

Catalog number: 380645000
500 mL, Plastic bottle, Each
Quantity
Packaging
Catalog number: 380645000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9005-84-9, 7732-18-5
Appearance (Form)
Clear to slightly hazy solution
Sensitivity
Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text