Borane-diphenylphosphine Complex, 94%, Thermo Scientific Chemicals
Borane-diphenylphosphine Complex, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Borane-diphenylphosphine Complex, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 380750010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namediphenylphosphane borane
Molecular FormulaC12H14BP
InChI KeyXUFGOLYVDOBYLK-UHFFFAOYSA-N
SMILESB.P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)200.03
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Elemental Analysis>=92.0 % (ex C)
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only