4-bromo-1-fluoro-2-nitrobenzene, 98%, Thermo Scientific Chemicals
4-bromo-1-fluoro-2-nitrobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-bromo-1-fluoro-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 380890250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-bromo-1-fluoro-2-nitrobenzene
Molecular FormulaC6H3BrFNO2
InChI KeyUQEANKGXXSENNF-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(Br)=CC=C1F
Molecular Weight (g/mol)220.00
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid after melting
Refractive index1.5750 to 1.5780 (20°C, 589 nm)
Appearance (Color)Clear yellow to brown
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only