tert-Butyl propargyl ether, 97%, Thermo Scientific Chemicals
tert-Butyl propargyl ether, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl propargyl ether, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 380910010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16314-18-4
IUPAC Name3-(tert-butoxy)prop-1-yne
Molecular FormulaC7H12O
InChI KeyQIGSPXPODKLGRK-UHFFFAOYSA-N
SMILESCC(C)(C)OCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4170 to 1.4190 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only