5-Bromo-1-methylimidazole, 97%, Thermo Scientific Chemicals
5-Bromo-1-methylimidazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-1-methylimidazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 381040250
Price (USD)/ Each
500.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
500.00
Add to cart
5-Bromo-1-methylimidazole, 97%, Thermo Scientific Chemicals
Catalog number381040250
Price (USD)/ Each
500.00
-
Add to cart
Chemical Identifiers
IUPAC Name5-bromo-1-methyl-1H-imidazole
Molecular FormulaC4H5BrN2
InChI KeyHATLLUIOEIXWGD-UHFFFAOYSA-N
SMILESCN1C=NC=C1Br
Molecular Weight (g/mol)161.00
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
Appearance (Form)Crystalline solid
Infrared spectrumConforms
Melting point38°C to 43°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only