Diethylaluminium iodide, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Diethylaluminium iodide, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylaluminium iodide, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 mL
Catalog number 381161000
Price (USD)/ Each
220.00
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
220.00
Add to cart
Diethylaluminium iodide, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Catalog number381161000
Price (USD)/ Each
220.00
-
Add to cart
Chemical Identifiers
CAS2040-00-8
IUPAC Namediethyl(iodo)alumane
Molecular FormulaC4H10AlI
InChI KeyPPQUYYAZSOKTQD-UHFFFAOYSA-M
SMILESCC[Al](I)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Strength19.5 to 20.5 wt % (on Al basis)
Appearance (Color)Clear colorless
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only