Thermo Scientific Chemicals

Triethylaluminium, 0.6M solution in heptane, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 381171000
100 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

Triethylaluminium, 0.6M solution in heptane, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 381171000
100 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 381171000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-93-8
IUPAC Name
triethylalumane
Molecular Formula
C6H15Al
InChI Key
VOITXYVAKOUIBA-UHFFFAOYSA-N
SMILES
CC[Al](CC)CC
Appearance (Color)
Clear colorless
Appearance (Form)
Solution
Strength
9.5 to 10.5 wt % (on Al basis)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text