n-Butylethylmagnesium, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
n-Butylethylmagnesium, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Butylethylmagnesium, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 381188000
Price (USD)/ Each
566.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
666.00 
Save 99.35 (15%)
-
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
566.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
666.00 
Save 99.35 (15%)
Add to cart
n-Butylethylmagnesium, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Catalog number381188000
Price (USD)/ Each
566.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
666.00 
Save 99.35 (15%)
-
Add to cart
Chemical Identifiers
CAS62202-86-2
IUPAC Namebutyl(ethyl)magnesium
Molecular FormulaC6H14Mg
InChI KeyMVECFARLYQAUNR-UHFFFAOYSA-N
SMILESCCCC[Mg]CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly hazy colorless
Titration13.5 to 14.5 % (on Mg basis)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only