Thermo Scientific Chemicals

Tris(trimethylsilyl) borate, Thermo Scientific Chemicals

Catalog number: 381240250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tris(trimethylsilyl) borate, Thermo Scientific Chemicals

Catalog number: 381240250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 381240250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4325-85-3
IUPAC Name
tritrimethylsilyl borate
Molecular Formula
C9H27BO3Si3
InChI Key
YZYKZHPNRDIPFA-UHFFFAOYSA-N
SMILES
C[Si](C)(C)OB(O[Si](C)(C)C)O[Si](C)(C)C
Appearance (Color)
Clear colorless
Refractive index
1.3850 to 1.3870 (20°C, 589 nm)
GC
>=97.0 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text