Thermo Scientific Chemicals

(tert-Butyldimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals

Catalog number: 381300010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(tert-Butyldimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals

Catalog number: 381300010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 381300010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86318-61-8
IUPAC Name
tert-butyl(ethynyl)dimethylsilane
Molecular Formula
C8H16Si
InChI Key
RTYNRTUKJVYEIE-UHFFFAOYSA-N
SMILES
CC(C)(C)[Si](C)(C)C#C
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless
GC
>=96.0 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text