Thermo Scientific Chemicals

Cyclopentylacetylene, 95%, Thermo Scientific Chemicals

Catalog number: 381310010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclopentylacetylene, 95%, Thermo Scientific Chemicals

Catalog number: 381310010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 381310010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
930-51-8
IUPAC Name
ethynylcyclopentane
Molecular Formula
C7H10
InChI Key
TXVJSWLZYQMWPC-UHFFFAOYSA-N
SMILES
C#CC1CCCC1
Appearance (Color)
Clear yellow
Refractive index
1.4470 to 1.4490 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text