Thermo Scientific Chemicals

Piperine, 98%, Thermo Scientific Chemicals

Catalog number: 381450250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Piperine, 98%, Thermo Scientific Chemicals

Catalog number: 381450250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 381450250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
94-62-2
IUPAC Name
(2E,4E)-5-(2H-1,3-benzodioxol-5-yl)-1-(piperidin-1-yl)penta-2,4-dien-1-one
Molecular Formula
C17H19NO3
InChI Key
MXXWOMGUGJBKIW-YPCIICBESA-N
SMILES
O=C(\C=C\C=C\C1=CC=C2OCOC2=C1)N1CCCCC1
Appearance (Color)
Off-white to yellow to tan
Infrared spectrum
Conforms
Melting point
128°C to 132°C
HPLC
>=97.5 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text