Thermo Scientific Chemicals

Trimethylsulfoxonium chloride, 97%, Thermo Scientific Chemicals

Catalog number: 381520010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Trimethylsulfoxonium chloride, 97%, Thermo Scientific Chemicals

Catalog number: 381520010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 381520010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5034-06-0
IUPAC Name
trimethyl(oxo)-λ⁶-sulfanylium chloride
Molecular Formula
C3H9ClOS
InChI Key
KQYWHJICYXXDSQ-UHFFFAOYSA-M
SMILES
[Cl-].C[S+](C)(C)=O
Appearance (Color)
White
Infrared spectrum
Conforms
Appearance (Form)
Powder
Titration Argentometric
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text