Trimethylsulfoxonium chloride, 97%, Thermo Scientific Chemicals
Trimethylsulfoxonium chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethylsulfoxonium chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 381520050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametrimethyl(oxo)-λ⁶-sulfanylium chloride
Molecular FormulaC3H9ClOS
InChI KeyKQYWHJICYXXDSQ-UHFFFAOYSA-M
SMILES[Cl-].C[S+](C)(C)=O
Molecular Weight (g/mol)128.61
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Titration Argentometric>=96.0 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only