Thermo Scientific Chemicals

4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: 382080250
25 g, Each
Thermo Scientific Chemicals

4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: 382080250
25 g, Each
Quantity
Catalog number: 382080250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3731-53-1
IUPAC Name
1-(pyridin-4-yl)methanamine
Molecular Formula
C6H8N2
InChI Key
TXQWFIVRZNOPCK-UHFFFAOYSA-N
SMILES
NCC1=CC=NC=C1
GC
>=97.5 %
Appearance (Color)
Clear colorless to yellow to orange
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.5500 to 1.5530 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text