Bis(norbornadiene)rhodium(I) tetrafluoroborate, 94%, Thermo Scientific Chemicals
Bis(norbornadiene)rhodium(I) tetrafluoroborate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(norbornadiene)rhodium(I) tetrafluoroborate, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Packaging:
Glass bottle
Catalog number 382555000
Price (USD)/ Each
-
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36620-11-8
IUPAC Nameλ¹-rhodium(1+) bis(bicyclo[2.2.1]hepta-2,5-diene) tetrafluoroboranuide
Molecular FormulaC14H16BF4Rh
InChI KeyHAYDJWBQWOEERB-UHFFFAOYSA-N
SMILES[Rh+].F[B-](F)(F)F.C1C2C=CC1C=C2.C1C2C=CC1C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
Appearance (Form)Powder
Infrared spectrumConforms
Elemental Analysis>=92 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only