Acetylacetonato(norbornadiene)rhodium(I), Thermo Scientific Chemicals
Acetylacetonato(norbornadiene)rhodium(I), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetylacetonato(norbornadiene)rhodium(I), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Packaging:
Glass bottle
Catalog number 382565000
Price (USD)/ Each
432.00
-
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
432.00
Add to cart
Acetylacetonato(norbornadiene)rhodium(I), Thermo Scientific Chemicals
Catalog number382565000
Price (USD)/ Each
432.00
-
Add to cart
Chemical Identifiers
CAS32354-50-0
Molecular FormulaC12H15O2Rh
Molecular Weight (g/mol)294.16
Synonymλ¹-rhodium(1+) norbornadiene (2Z)-4-oxopent-2-en-2-olate
MDL NumberMFCD00075071
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Powder
Infrared spectrumConforms
Rhodium (Rh)>=34.7 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only