Specifications
Chemical Name or MaterialSodium phosphate dibasic
Melting Point250°C
CAS7558-79-4
ColorWhite
Quantity12 kg
Molecular FormulaNa2HPO4
SynonymDisodium hydrogen orthophosphate, Sodium hydrogen phosphate
Molecular Weight (g/mol)141.96
Percent Purity≥98%
GradeACS/USP Volumetric
PackagingPoly pail, screw top
Unit SizeEach