Thionyl chloride, 99.7%, Thermo Scientific Chemicals
Thionyl chloride, 99.7%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thionyl chloride, 99.7%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 382660010
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7719-09-7
IUPAC Namesulfurooyl dichloride
Molecular FormulaCl2OS
InChI KeyFYSNRJHAOHDILO-UHFFFAOYSA-N
SMILESClS(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Color scale=<50 APHA
Infrared spectrumConforms
GC>=99.6 % (SOCl2)
Impurity=<0.3 % (SO2)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection