Thermo Scientific Chemicals

(R)-(+)-3-Aminoquinuclidine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 382890010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(R)-(+)-3-Aminoquinuclidine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 382890010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 382890010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
123536-14-1
IUPAC Name
1-azabicyclo[2.2.2]octan-3-amine dihydrochloride
Molecular Formula
C7H16Cl2N2
InChI Key
STZHBULOYDCZET-UHFFFAOYNA-N
SMILES
Cl.Cl.NC1CN2CCC1CC2
Appearance (Color)
White to beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
Specific optical rotation
+22° to +26° (20°C, 589 nm) (C=1, Water)
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text