2-(Diphenylphosphino)benzoic Acid, 98%, Thermo Scientific Chemicals
2-(Diphenylphosphino)benzoic Acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Diphenylphosphino)benzoic Acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 382930010
Price (USD)/ Each
73.60
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
73.60
Add to cart
2-(Diphenylphosphino)benzoic Acid, 98%, Thermo Scientific Chemicals
Catalog number382930010
Price (USD)/ Each
73.60
-
Add to cart
Chemical Identifiers
CAS17261-28-8
IUPAC Name2-(diphenylphosphanyl)benzoic acid
Molecular FormulaC19H15O2P
InChI KeyUYRPRYSDOVYCOU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Melting point177°C to 181°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only