Thermo Scientific Chemicals

Bis(dichlorophosphino)methane, 96%, Thermo Scientific Chemicals

Catalog number: 383000010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Bis(dichlorophosphino)methane, 96%, Thermo Scientific Chemicals

Catalog number: 383000010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 383000010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28240-68-8
IUPAC Name
dichloro[(dichlorophosphanyl)methyl]phosphane
Molecular Formula
CH2Cl4P2
InChI Key
OEKLCAXLANZKPX-UHFFFAOYSA-N
SMILES
ClP(Cl)CP(Cl)Cl
NMR (1H-NMR)
Consistent with structure
Appearance (Color)
Clear colorless to light yellow
Titration Argentometric
>=95 %
Appearance (Form)
Liquid
Refractive index
1.5920 to 1.5950 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text