Diglycidyl ether, 95%, Thermo Scientific Chemicals
Diglycidyl ether, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diglycidyl ether, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 383130010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2238-07-5
IUPAC Name2-{[(oxiran-2-yl)methoxy]methyl}oxirane
Molecular FormulaC6H10O3
InChI KeyGYZLOYUZLJXAJU-UHFFFAOYNA-N
SMILESC(OCC1CO1)C1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to brown
Infrared spectrumConforms
Refractive index1.4460 to 1.4500 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only