Aluminum trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals
Aluminum trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Plastic bottle
Catalog number 383260500
Price (USD)/ Each
272.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
320.00 
Save 47.35 (15%)
-
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
272.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
320.00 
Save 47.35 (15%)
Add to cart
Aluminum trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals
Catalog number383260500
Price (USD)/ Each
272.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
320.00 
Save 47.35 (15%)
-
Add to cart
Chemical Identifiers
CAS74974-61-1
IUPAC Namealuminium(3+) tritrifluoromethanesulfonate
Molecular FormulaC3AlF9O9S3
InChI KeyFKOASGGZYSYPBI-UHFFFAOYSA-K
SMILES[Al+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Complexometric>=98.5 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only